ENA447957277 Enamine

1-{1-phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl}piperidine

  • MDL number MFCD00420078
  • Cas Number : 23000-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206573
Availability details for
Quantity
  • Availability
  • Add to Cart