ENA447957280 Enamine

2-(3,4-difluorophenyl)-N-(thiophen-3-yl)morpholine-4-carboxamide

  • MDL number MFCD28383985
  • Cas Number : 1803601-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206577
Availability details for
Quantity
  • Availability
  • Add to Cart