ENA447957283 Enamine

ethyl 2-(chlorosulfonyl)pentanoate

  • MDL number MFCD24358382
  • Cas Number : 1556927-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206589
Availability details for
Quantity
  • Availability
  • Add to Cart