ENA447957302 Enamine

7-bromoimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09995206
  • Cas Number : 1019018-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206662
Availability details for
Quantity
  • Availability
  • Add to Cart