ENA447957305 Enamine

5-methyl-1-[(5-methyl-1,2-oxazol-3-yl)methyl]-1H-1,2,3-triazol-4-amine

  • MDL number MFCD21947807
  • Cas Number : 1518746-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206671
Availability details for
Quantity
  • Availability
  • Add to Cart