ENA447957312 Enamine

3-(benzyloxy)propane-1-sulfonamide

  • MDL number MFCD19676414
  • Cas Number : 1408646-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206684
Availability details for
Quantity
  • Availability
  • Add to Cart