ENA447957316 Enamine

5-amino-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00178662
  • Cas Number : 52217-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206690
Availability details for
Quantity
  • Availability
  • Add to Cart