ENA447957320 Enamine

4-bromo-2-fluoro-N-[2-(pyrrolidin-1-yl)ethyl]benzamide

  • MDL number MFCD13197720
  • Cas Number : 1307443-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206696
Availability details for
Quantity
  • Availability
  • Add to Cart