ENA447957322 Enamine

butyl methyl carbonate

  • MDL number MFCD19105215
  • Cas Number : 4824-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206705
Availability details for
Quantity
  • Availability
  • Add to Cart