ENA447957330 Enamine

1,4-dichloro-4-methylpentane

  • MDL number MFCD00204041
  • Cas Number : 54305-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205464
Availability details for
Quantity
  • Availability
  • Add to Cart