ENA447957333 Enamine

methyl 5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD17324128
  • Cas Number : 1275687-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206760
Availability details for
Quantity
  • Availability
  • Add to Cart