ENA447957342 Enamine

3-(benzyloxy)propane-1-sulfonyl chloride

  • MDL number MFCD19669372
  • Cas Number : 1498463-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206796
Availability details for
Quantity
  • Availability
  • Add to Cart