ENA447957343 Enamine

4-(piperazin-1-yl)quinoline dihydrochloride

  • MDL number MFCD26383403
  • Cas Number : 1350361-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206797
Availability details for
Quantity
  • Availability
  • Add to Cart