ENA447957348 Enamine

2-[3-(2-methylpropyl)phenyl]propanoic acid

  • MDL number MFCD08275582
  • Cas Number : 66622-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206804
Availability details for
Quantity
  • Availability
  • Add to Cart