ENA447957352 Enamine

5-bromo-1,2-dimethyl-1H-1,3-benzodiazole

  • MDL number MFCD16250432
  • Cas Number : 99513-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206808
Availability details for
Quantity
  • Availability
  • Add to Cart