ENA447957353 Enamine

1-isothiocyanato-1-methylcyclopropane

  • MDL number MFCD02684164
  • Cas Number : 1028018-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205111
Availability details for
Quantity
  • Availability
  • Add to Cart