ENA447957354 Enamine

4-(4-ethylpiperazin-1-yl)-2-methoxyaniline

  • MDL number MFCD18605332
  • Cas Number : 761440-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206810
Availability details for
Quantity
  • Availability
  • Add to Cart