ENA447957356 Enamine

ethyl 2-chloro-5-(chlorosulfonyl)-4-fluorobenzoate

  • MDL number MFCD12169528
  • Cas Number : 1155076-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206812
Availability details for
Quantity
  • Availability
  • Add to Cart