ENA447957359 Enamine

1-[(5-chloro-2-methoxyphenyl)carbamoyl]azetidine-3-carboxylic acid

  • MDL number MFCD22749413
  • Cas Number : 1492395-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206820
Availability details for
Quantity
  • Availability
  • Add to Cart