ENA447957365 Enamine

5,6-dichloro-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD24562188
  • Cas Number : 110568-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206845
Availability details for
Quantity
  • Availability
  • Add to Cart