ENA447957366 Enamine

[5-(2-methoxyphenyl)-1H-pyrazol-3-yl]methanol

  • MDL number MFCD16305649
  • Cas Number : 939053-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206846
Availability details for
Quantity
  • Availability
  • Add to Cart