ENA447957383 Enamine

3-bromo-5-methyl-1,4-dihydropyridin-4-one

  • MDL number MFCD20673647
  • Cas Number : 70149-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206867
Availability details for
Quantity
  • Availability
  • Add to Cart