ENA447957384 Enamine

2-(2-methoxyphenyl)-5-methylpiperidine

  • MDL number MFCD20311903
  • Cas Number : 1339159-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206868
Availability details for
Quantity
  • Availability
  • Add to Cart