ENA447957385 Enamine

tert-butyl 1-oxo-1lambda4-thiomorpholine-4-carboxylate

  • MDL number MFCD21140584
  • Cas Number : 278788-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206869
Availability details for
Quantity
  • Availability
  • Add to Cart