ENA447957387 Enamine

2',3'-dihydro-1'H-spiro[cyclobutane-1,4'-isoquinoline]

  • MDL number MFCD16294407
  • Cas Number : 850875-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206871
Availability details for
Quantity
  • Availability
  • Add to Cart