ENA447957388 Enamine

5-chloro-2-fluorobenzene-1-thiol

  • MDL number MFCD12026160
  • Cas Number : 1208076-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206872
Availability details for
Quantity
  • Availability
  • Add to Cart