ENA447957398 Enamine

ethyl 4-aminocyclohexane-1-carboxylate hydrochloride

  • MDL number MFCD12913688
  • Cas Number : 90950-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206920
Availability details for
Quantity
  • Availability
  • Add to Cart