ENA447957400 Enamine

3-(3-aminophenyl)-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD14640866
  • Cas Number : 842129-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205484
Availability details for
Quantity
  • Availability
  • Add to Cart