ENA447957402 Enamine

6-amino-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD07369810
  • Cas Number : 186774-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206942
Availability details for
Quantity
  • Availability
  • Add to Cart