ENA447957403 Enamine

2,2-dimethyl-6-nitro-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD07369941
  • Cas Number : 111478-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206943
Availability details for
Quantity
  • Availability
  • Add to Cart