ENA447957406 Enamine

ethyl 2-[2-(2-methyl-4-nitrophenyl)hydrazin-1-ylidene]propanoate

  • Cas Number : 93185-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207009
Availability details for
Quantity
  • Availability
  • Add to Cart