ENA447957407 Enamine

2-methyl-4-propyl-1,3-thiazole

  • MDL number MFCD26797022
  • Cas Number : 41981-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207017
Availability details for
Quantity
  • Availability
  • Add to Cart