ENA447957410 Enamine

1-benzoylpiperidin-2-one

  • MDL number MFCD18083538
  • Cas Number : 4252-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207021
Availability details for
Quantity
  • Availability
  • Add to Cart