ENA447957411 Enamine

ethyl 2-methyl-2-(piperidin-4-yl)propanoate hydrochloride

  • MDL number MFCD08703276
  • Cas Number : 243836-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207022
Availability details for
Quantity
  • Availability
  • Add to Cart