ENA447957413 Enamine

2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methoxypentanoic acid

  • MDL number MFCD23724127
  • Cas Number : 1483389-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207025
Availability details for
Quantity
  • Availability
  • Add to Cart