ENA447957420 Enamine

2-(1H-pyrrol-1-yl)pyridine-3-carboxylic acid

  • MDL number MFCD18855239
  • Cas Number : 154014-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207032
Availability details for
Quantity
  • Availability
  • Add to Cart