ENA447957422 Enamine

methyl 2-(piperidin-1-yl)benzoate

  • MDL number MFCD01569808
  • Cas Number : 81215-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207035
Availability details for
Quantity
  • Availability
  • Add to Cart