ENA447957427 Enamine

2-(chloromethyl)-4-(4-methylphenyl)-1,3-thiazole

  • MDL number MFCD08060632
  • Cas Number : 921102-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207053
Availability details for
Quantity
  • Availability
  • Add to Cart