ENA447957430 Enamine

N2-ethyl-1,3,5-triazine-2,4-diamine

  • MDL number MFCD19204121
  • Cas Number : 30368-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-197
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207056
Availability details for
Quantity
  • Availability
  • Add to Cart