ENA447957435 Enamine

ethyl 2-[(dimethylsulfamoyl)amino]acetate

  • MDL number MFCD02861217
  • Cas Number : 163078-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207064
Availability details for
Quantity
  • Availability
  • Add to Cart