ENA447957437 Enamine

[(2-methoxyethyl)sulfamoyl]dimethylamine

  • MDL number MFCD02860427
  • Cas Number : 349420-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207067
Availability details for
Quantity
  • Availability
  • Add to Cart