ENA447957438 Enamine

4,4-dimethyl-1-(1-methyl-1H-pyrazol-3-yl)pentan-1-amine

  • MDL number MFCD21131774
  • Cas Number : 1496196-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207068
Availability details for
Quantity
  • Availability
  • Add to Cart