ENA447957441 Enamine

4-bromo-3-cyclopropyl-1H-pyrazol-5-ol

  • MDL number MFCD23843037
  • Cas Number : 1808396-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207078
Availability details for
Quantity
  • Availability
  • Add to Cart