ENA447957442 Enamine

1-(3-chlorophenyl)-2-hydroxypropan-1-one

  • MDL number MFCD16165818
  • Cas Number : 152943-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205498
Availability details for
Quantity
  • Availability
  • Add to Cart