ENA447957446 Enamine

methyl 5-chloro-6-cyanopyridine-2-carboxylate

  • MDL number MFCD23706794
  • Cas Number : 1427392-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205517
Availability details for
Quantity
  • Availability
  • Add to Cart