ENA447957459 Enamine

7-acetyl-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD06252057
  • Cas Number : 84330-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-198
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207113
Availability details for
Quantity
  • Availability
  • Add to Cart