ENA447957461 Enamine

1-[2-(oxan-2-yloxy)ethyl]piperidin-3-amine

  • MDL number MFCD14643702
  • Cas Number : 1247144-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205540
Availability details for
Quantity
  • Availability
  • Add to Cart