ENA447957463 Enamine

1-(4-amino-3-hydroxyphenyl)ethan-1-one

  • MDL number MFCD01727691
  • Cas Number : 54903-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207119
Availability details for
Quantity
  • Availability
  • Add to Cart