ENA447957465 Enamine

3-fluoro-1-iodobutane

  • MDL number MFCD24462490
  • Cas Number : 7411-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207121
Availability details for
Quantity
  • Availability
  • Add to Cart