ENA447957469 Enamine

N-[2-(benzyloxy)phenyl]acetamide

  • MDL number MFCD03090753
  • Cas Number : 34288-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207125
Availability details for
Quantity
  • Availability
  • Add to Cart